Learn More
Avantor™ BAKERBOND Speedisk™ Extraction Disks
Maximize capacity, adsorption and SPE speed. Avantor BAKERBOND Speedisk Extraction Disks are neither cartridge nor membrane that provides an excellent SPE tool for environmental samples and combines a thin bed of BAKERBOND sorbent with a special laminar configuration for testing drinking water, ground water, waste water, solid waste or process streams.
$4451.67 - $5317.33
Specifications
For Use With (Equipment) Expanded Extraction Station (8095-06)
Sorbent C18 XF
Quantity 20/Pk.
Diameter (Metric) Outer 50 mm
View More Specs
Products 2
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
02002377
View Documents
Avantor™
805606
Octadecyl (C18 XF) Extraction Disk
Each for $5,317.33
Only null left
02002378
View Documents
Avantor™
805706
Octyl (C8) Extraction Disk
Each for $4,451.67
Only null left
Description
Description
  • Pre-assembled with rigid support and baffle to maximize laminar flow, allow for filtration, resist clogging, and maximize capacity, adsorption, and speed
  • Require minimal glassware, set up time, and assembly
  • Minimizes impact of extractable compounds
  • Resists clogging and ensures high throughput rate
  • Reduce SPE run times
Specifications
Specifications
Expanded Extraction Station (8095-06)
C18 XF
20/Pk.
50 mm
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'