Learn More
Aushon Chemiluminescent Human Cytokine Array Kits
For simultaneous measurement of combinations of cytokines
Supplier: Aushon Biosystems Inc 020071FA
Description
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating cytokine profiles
Greener Choice Claims
- Environmental benefits include:
- Non-Toxic
- Recyclable
Specifications
Human | |
For simultaneous measurement of combinations of cytokines | |
Cytokine Array Kits | |
3 Plex |