missing translation for 'onlineSavingsMsg'
Learn More

Arsenic standard solution, for AAS, 1 mg/ml As in 2% KOH

CAS: 7440-38-2 | As | 74.92 g/mol

Supplier:  Thermo Scientific Chemicals 195825000

 View more versions of this product

Catalog No. AC195825000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for atomic adsorption spectroscopy
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications

Specifications

Arsenic standard solution
Plastic bottle
1.02g/mL
As
MFCD00085309
RQNWIZPPADIBDY-UHFFFAOYSA-N
arsenic
500 mL
CHEBI:27563
AAS
1 mg/ml As in 2% KOH
7440-38-2,1310-58-3,7732-18-5
Solution
997 to 1003 μg/mL of As(3+) in 2% KOH (20°C)
15, 784
[As]
74.92
5359596
74.92
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only