missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arsenic III Speciation Standard (Assurance⢠Grade/For ICP-MS), SPEX CertiPrepā¢
$127.17 - $127.17
Chemical Identifiers
CAS | 7440-38-2 |
---|---|
Molecular Formula | As |
Molecular Weight (g/mol) | 74.92 |
MDL Number | MFCD00085309 |
InChI Key | RQNWIZPPADIBDY-UHFFFAOYSA-N |
PubChem CID | 5359596 |
ChEBI | CHEBI:27563 |
IUPAC Name | arsenic |
SMILES | [As] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
SPECAS3M
|
SPEX CertiPrep Group LLC
SPECAS3M |
30 mL | Triple-leached LDPE bottle |
Each for $127.17
|
|
|||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
As(+3) | 1000μg/mL |
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Chemical Identifiers
7440-38-2 | |
74.92 | |
RQNWIZPPADIBDY-UHFFFAOYSA-N | |
CHEBI:27563 | |
[As] |
As | |
MFCD00085309 | |
5359596 | |
arsenic |
Specifications
Triple-leached LDPE bottle | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
0.1,2.0,97.9 | |
MFCD00085309 | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
ICP/ICP-MS Standard | |
[As] | |
arsenic | |
30 mL | |
CHEBI:27563 | |
Arsenic III Speciation Standard |
7440-38-2,7647-01-0,7732-18-5 | |
0.1,2.0,97.9 | |
As | |
ICP-MS | |
Standard Solution | |
RQNWIZPPADIBDY-UHFFFAOYSA-N | |
2% HCl | |
74.92 | |
5359596 | |
Assurance |
Safety and Handling
Causes skin irritation, serious eye irritation
DOTInformation : Nonhazardous
Recommended Storage : Ambient