Learn More
Applied Biosystems™ GeneAmp™ dNTP Blend (100 mM)
GeneAmp dNTPs are deoxynucleotide triphosphates dissolved in glass-distilled water and titrated to pH 7.0 with NaOH. Each vial contains a blend of 100 mM solution of dNTP (25 mM each of dATP, dCTP, dGTP and dTTP).
$1,452.00
Specifications
| Concentration | 100 mM |
|---|---|
| Form | Liquid |
| Product Line | GeneAmp |
| Shipping Condition | Dry Ice |
| For Use With (Application) | Standard PCR |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
N8080261
|
Applied Biosystems™
N8080261 |
1 mL |
Each for $1,452.00
|
|
|||||
4303443
|
Applied Biosystems™
4303443 |
12 x 1 mL |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
GeneAmp dNTPs are deoxynucleotide triphosphates dissolved in glass-distilled water and titrated to pH 7.0 with NaOH. Each vial contains a blend of 100 mM solution of dNTP (25 mM each of dATP, dCTP, dGTP and dTTP). Sufficient for 2000 PCR amplifications, 50 µL each. Each kit contains 12 x 1 mL vials.
Key advantages of GeneAmp dNTPs
• High purity dNTPs are ideal for use in PCR reactions or other applications
• Guaranteed for PCR performance when used with GeneAmp PCR core reagents and GeneAmp instrument systems
• Premixed dNTP blends are available in various concentrations, offering ease of use
Specifications
| 100 mM | |
| GeneAmp | |
| Standard PCR |
| Liquid | |
| Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.