missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneAmp™ dNTP Blend (100 mM)

Catalog No. 4303443
Encompass
Change view
Click to view available options
Quantity:
1 mL
12 x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4303443 12 x 1 mL
N8080261 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4303443 Supplier Applied Biosystems™ Supplier No. 4303443

GeneAmp dNTPs are deoxynucleotide triphosphates dissolved in glass-distilled water and titrated to pH 7.0 with NaOH. Each vial contains a blend of 100 mM solution of dNTP (25 mM each of dATP, dCTP, dGTP and dTTP).

GeneAmp dNTPs are deoxynucleotide triphosphates dissolved in glass-distilled water and titrated to pH 7.0 with NaOH. Each vial contains a blend of 100 mM solution of dNTP (25 mM each of dATP, dCTP, dGTP and dTTP). Sufficient for 2000 PCR amplifications, 50 µL each. Each kit contains 12 x 1 mL vials.

Key advantages of GeneAmp dNTPs
• High purity dNTPs are ideal for use in PCR reactions or other applications
• Guaranteed for PCR performance when used with GeneAmp PCR core reagents and GeneAmp instrument systems
• Premixed dNTP blends are available in various concentrations, offering ease of use

Specifications

Concentration 100 mM
Content And Storage GeneAmp dNTP Blend, 100 mM (12 x 1 mL)

Store in a constant temperature freezer at –20°C.
Form Liquid
Product Line GeneAmp
Quantity 12 x 1 mL
Shipping Condition Dry Ice
For Use With (Application) Standard PCR
Why do some dNTP mixes contain dTTP and others TTP?

Thymine is normally found as a deoxyribose. Thus, TTP and dTTP are essentially the same thing. Both types of mixes can be used in PCR reactions

For Research Use Only. Not for use in diagnostic procedures.