missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP2 alpha Monoclonal Antibody (K92029_16H5), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Fisher Chemical MA555721
Description
Sequence of this protein is as follows: EDLLHGPHAL SSGLGDLSIH SLPHAIEEVP HVEDPGINIP DQTVIKKGPV SLSKSNSNAV SAIPINKDNL FGGVVNPNEV FCSVPGRLSL LSSTSKYKVT VAEVQRRLSP PECLNASLLG GVLRRAKSKN GGRSLREKLD KIGLNLPAGR RKAANVTLLT SLVEGEAVHL ARDFGYVCET EFPAKAVAEF LNRQHSDPNE QVTRKNMLLA TKQICKEFTD LLAQDRSPLG NSRPNPILEP GIQSCLTHFN LISHGFGSPA VCAAVTALQN YLTEALKAMD KMYLSNNPNS HTDNNAKSSD
AP2 alpha Monoclonal antibody specifically detects AP2 alpha in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
AP2 alpha | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P05549 | |
Mouse | |
Protein A | |
RUO | |
7020 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunoprecipitation, Western Blot | |
K92029_16H5 | |
Unconjugated | |
TFAP2A | |
activating enhancer-binding protein 2 alpha; activating enhancer-binding protein 2-alpha; Activator protein 2; activator protein 2 alpha variant 4; activator protein 2 alpha variant 6; activator protein 2 alpha variant 7; AP2; AP-2; Ap-2 (a); AP-2 transcription factor; AP2alpha; AP-2alpha; AP2-alpha; Ap2tf; BOFS; ERF-1; FLJ51761; OTTHUMP00000214240; OTTHUMP00000214243; Tcfap2a; TFAP2; TFAP2A; transcription factor AP2; transcription factor AP-2 alpha; transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha); transcription factor AP-2, alpha; transcription factor AP-2-alpha; transcription factor AP-2-alpha-like; transcription factor; AP-2 | |
Recombinant human TFAP2A (immunogen range is 131-430). | |
50 ÎĽg | |
Primary | |
Human | |
Antibody | |
IgG2a |