missing translation for 'onlineSavingsMsg'
Learn More
Learn More
thymosin beta 4, X-linked, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human TMSB4X protein.
Supplier: Abnova Corporation H00007114B01P
Description
This gene encodes an actin sequestering protein which plays a role in regulation of actin polymerization. The protein is also involved in cell proliferation, migration, and differentiation. This gene escapes X inactivation and has a homolog on chromosome Y. [provided by RefSeq
Sequence: MSDKPDMAEIEKFDKSKLKKTETQEKNPLPSKETIEQEKQAGESGreener Choice Claims
- Environmental benefits include:
- Recyclable
Specifications
thymosin beta 4, X-linked | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human TMSB4X protein. | |
TMSB4X | |
FX/PTMB4/TB4X/TMSB4 | |
Mouse | |
Affinity Purified | |
RUO | |
7114 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Immunofluorescence, Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC001631 | |
TMSB4X | |
TMSB4X (AAH01631.1, 1 a.a. ∼ 44 a.a) full-length human protein. | |
50 μg | |
Primary | |
Human | |
Antibody |