missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prospero homeobox protein 1/PROX1 Rabbit anti-Rat, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABN278
Description
This Anti-Prospero homeobox protein 1/PROX1 Antibody is validated for use in western blotting and immunohistochemistry for the detection of Prospero homeobox protein 1/PROX1.Specifications
Prospero homeobox protein 1/PROX1 | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Mouse, Rat |
Immunohistochemistry, Western Blot | |
Unconjugated | |
PROX1 | |
KLH-conjugated linear peptide corresponding to the C-terminus of human Prospero homeobox protein 1/PROX1. | |
100 μL | |
NP_001257545 | |
2-8°C for 12 months |
For Research Use Only