missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metabotropic Glutamate Receptor 7 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB9213
Description
Detect Metabotropic Glutamate Receptor 7 using this Anti-Metabotropic Glutamate Receptor 7 Antibody validated for use in immunohistochemistry (paraffin) and western blotting.Specifications
Metabotropic Glutamate Receptor 7 | |
Polyclonal | |
Affinity purified immunoglobulin. Liquid in PBS 0.01% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
GRM7; mGlu7; GPRC1G; FLJ40498; mGluR7; MGLUR7; GLUR7 | |
Synthetic peptide from the N-terminal extracellular domain of human Metabotropic Glutamate Receptor 7. The immunogen has 100% homology with rat. | |
50 μg | |
NM_181875.1; NM_000844.2; NM_181874.1 | |
−70°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles. |
For Research Use Only