missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin αM [CD11b] Mouse anti-Rat, Clone: OX-42, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma CBL1512
Description
Detect Integrin αM [CD11b] using this Anti-Integrin αM CD11b Antibody, clone OX-42 validated for use in flow cytometry, immunohistochemistry, immunohistochemistry (paraffin) and immunoprecipitation.Specifications
Integrin αM [CD11b] | |
Monoclonal | |
Unconjugated | |
NM_000632.3 | |
Mouse | |
Protein A purfied | |
Primary | |
4°C for up to 1 month, −20°C in undiluted aliquots for long term | |
IgG2a |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
OX-42 | |
The monoclonal is presented in phosphate buffered saline containing 10mM sodium azide. | |
ITGAM, MO1A, CD11b, MAC1A, CD11B, MAC-1, CR3A, MGC117044 | |
Rat peritoneal macrophages | |
500 μg | |
Rat | |
Purified |
For Research Use Only