missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza B Mouse anti-Human, Clone: B2 and B4 Blend, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB8661
Description
Anti-Influenza B Antibody, nucleoprotein, clones B2, B4 Blend detects level of Influenza B and has been published and validated for use in immunofluoresence.Specifications
Influenza B | |
Monoclonal | |
Unconjugated | |
Mouse | |
Unpurified | |
Primary | |
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG |
Immunofluorescence | |
B2 and B4 Blend | |
Ascites fluid with 0.1% sodium azide as a preservative. | |
Influenza Blend. | |
100 μL | |
Human | |
Ascites |
For Research Use Only