missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hypoxia Inducible Factor 1 α Mouse anti-Ferret, Human, Mouse, Rat, Clone: H1alpha67, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB5382
Description
Anti-Hypoxia Inducible Factor 1 α Antibody, clone H1α67 is a Mouse Monoclonal Antibody for detection of Hypoxia Inducible Factor 1 α also known as HIF-1 alpha or ARNT Interacting Protein and has been validated in EMSA, immunohistochemistry, immunoprecipitation and western blotting.Greener Choice Claims
- Environmental benefits include:
- Source Reduction
- Renewable Energy
Specifications
Hypoxia Inducible Factor 1 α | |
Monoclonal | |
Unconjugated | |
NM_001530.2; NM_181054.1 | |
Mouse | |
Protein A purfied | |
Primary | |
2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG2b |
Electromobility Shift Assay, Immunohistochemistry, Immunoprecipitation, Western Blot | |
H1alpha67 | |
Protein A Purified mouse immunoglobulin in 20mM sodium phosphate, 250mM NaCl, pH. 7.6, with 0.1% sodium azide as a preservative. | |
HIF1A, HIF-1ALPHA, HIF-1alpha, MOP1, PASD8, HIF1-ALPHA, HIF1 | |
Fusion protein from amino acids 432-528 of human HIF-1alpha. | |
100 μg | |
Ferret, Human, Mouse, Rat, Sheep | |
Purified |
For Research Use Only