missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-B27 Mouse anti-Human, Clone: HLA.ABC.m3, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB1285
Description
This Anti-HLA-B27 Antibody, clone HLA.ABC.m3 is validated for use in flow cytometry, IC for the detection of HLA-B27.Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
HLA-B27 | |
Monoclonal | |
Unconjugated | |
NM_005514.5 | |
Mouse | |
100 μg | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry | |
HLA.ABC.m3 | |
The antibody is supplied in phosphate-buffered saline, pH 7.4, containing 0.2% bovine serum albumin and 0.1% sodium azide. The characteristics of each lot are tested by flow cytometry. | |
HLA-B, Bw-54, Bw-45, Bw-47, Bw-46, Bw-52, B-5, Bw-4, Bw-57, OTTHUMP00000034826, AS, Bw-56, Bw-55, Bw-41, Bw-48, B' DT, Bw-50, Bw-60, Bw-58, B-21, HLA-B-5501, B-12, Bw-22, Bw-53, HLAB, SPDA1, Bw-44 | |
Immune complex precipitated from HLA-B27 positive cell line (Bordin) by anti HLA antibody and staphylococcal Protein A. | |
Primary | |
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG2a |
For Research Use Only