missing translation for 'onlineSavingsMsg'
Learn More
Learn More
chondroitin sulfate proteoglycan 5 (neuroglycan C), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CSPG5.
Supplier: Abnova Corporation H00010675A01
Description
Sequence: KKLYLLKTENTKLRRTNKFRTPSELHNDNFSLSTIAEGSHPNDDPSAPHKIQEVLKSCLKEEESFNIQNSMSPKLEGGKGDQADLDVNCLQNNLTGreener Choice Claims
- Environmental benefits include:
- Recyclable
Specifications
chondroitin sulfate proteoglycan 5 (neuroglycan C) | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant CSPG5. | |
CSPG5 | |
MGC44034/NGC | |
Mouse | |
50 μL | |
Neurobiology | |
Primary | |
Human | |
Serum |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_006574 | |
CSPG5 | |
CSPG5 (NP_006565, 445 a.a. ∼ 539 a.a) partial recombinant protein with GST tag. | |
RUO | |
Yes | |
10675 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |