Learn More
MilliporeSigma™ Annexin V-PE Apoptosis Detection Kit
Identifies phosphatidylserine (PS) that translocates from cytoplasmic face of plasma membrane to cell surface during apoptosis.
Supplier: MilliporeSigma™ CBA060100TEST
Description
- Has strong natural affinity for phosphatidylserine
- Performed on live cells
- Used for detecting apoptosis
Specifications
Annexin V-FITC, Binding Buffer, Propidium Iodide and user protocol | |
1 Kit |