Learn More
Ammonium hexachloroosmate(IV), Premion™, 99.99% (metals basis), Os 42.8% min
CAS: 12125-08-5 | Cl6H8N2Os | 439.00 g/mol
Supplier: Thermo Scientific Chemicals 01082303
Description
Ammonium hexachloroosmate(IV) is suitable for glass, optic and ceramic applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Environmental benefits include:
- Renewable Energy
Specifications
Ammonium hexachloroosmate(IV) | |
12125-08-5 | |
Powder | |
(metals basis) | |
(NH4)2OsCl6 | |
14,536 | |
Soluble in water. | |
[NH4+].[NH4+].Cl[Os+4](Cl)(Cl)(Cl)(Cl)Cl | |
439.00 | |
438.99 | |
Premion™ | |
Hygroscopic |
170°C (sublimation) | |
Brown to Black | |
1 g | |
Cl6H8N2Os | |
MFCD00010883 | |
ammonium hexachloroosmate iv, nh4 2oscl6, osmium iv-ammonium chloride, diazanium;hexachloroosmium 2-, diammonium hexachloroosmiumdiuide, ammonium hexachloroosmiate iv , premion | |
JRHMYOOMZKAPKT-UHFFFAOYSA-J | |
diazanium;hexachloroosmium(2-) | |
11729867 | |
99.99% | |
Os 42.8% min. | |
2.9 g/mL |
Chemical Identifiers
12125-08-5 | |
439.00 | |
JRHMYOOMZKAPKT-UHFFFAOYSA-J | |
11729867 |
Cl6H8N2Os | |
MFCD00010883 | |
ammonium hexachloroosmate iv, nh4 2oscl6, osmium iv-ammonium chloride, diazanium;hexachloroosmium 2-, diammonium hexachloroosmiumdiuide, ammonium hexachloroosmiate iv , premion | |
[NH4+].[NH4+].Cl[Os+4](Cl)(Cl)(Cl)(Cl)Cl |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 235-188-5
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only