missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexachloroosmate(IV), Premion™, 99.99% (metals basis), Os 42.8% min

CAS: 12125-08-5 | Cl6H8N2Os | 439.00 g/mol

$338.95 - $1221.59

Chemical Identifiers

CAS 12125-08-5
Molecular Formula Cl6H8N2Os
Molecular Weight (g/mol) 439.00
MDL Number MFCD00010883
InChI Key JRHMYOOMZKAPKT-UHFFFAOYSA-J
Synonym ammonium hexachloroosmate iv, nh4 2oscl6, osmium iv-ammonium chloride, diazanium;hexachloroosmium 2-, diammonium hexachloroosmiumdiuide, ammonium hexachloroosmiate iv , premion
PubChem CID 11729867
SMILES [NH4+].[NH4+].Cl[Os+4](Cl)(Cl)(Cl)(Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1082303
View Documents
Thermo Scientific Chemicals
01082303
1 g
Each for $338.95
Only null left
 
AA1082306
View Documents
Thermo Scientific Chemicals
01082306
5 g
Each for $1,221.59
Only null left
 
Description

Description

Ammonium hexachloroosmate(IV) is suitable for glass, optic and ceramic applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12125-08-5
439.00
JRHMYOOMZKAPKT-UHFFFAOYSA-J
11729867
Cl6H8N2Os
MFCD00010883
ammonium hexachloroosmate iv, nh4 2oscl6, osmium iv-ammonium chloride, diazanium;hexachloroosmium 2-, diammonium hexachloroosmiumdiuide, ammonium hexachloroosmiate iv , premion
[NH4+].[NH4+].Cl[Os+4](Cl)(Cl)(Cl)(Cl)Cl

Specifications

170°C (sublimation)
Brown to Black
1 g
(NH4)2OsCl6
14,536
Soluble in water.
[NH4+].[NH4+].Cl[Os+4](Cl)(Cl)(Cl)(Cl)Cl
439.00
438.99
Premion
Hygroscopic
Ammonium hexachloroosmate(IV)
12125-08-5
Powder
Cl6H8N2Os
MFCD00010883
ammonium hexachloroosmate iv, nh4 2oscl6, osmium iv-ammonium chloride, diazanium;hexachloroosmium 2-, diammonium hexachloroosmiumdiuide, ammonium hexachloroosmiate iv , premion
JRHMYOOMZKAPKT-UHFFFAOYSA-J
diazanium;hexachloroosmium(2-)
11729867
99.99%
Os 42.8% min.
2.9 g/mL
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 235-188-5

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only