Learn More
Ahlstrom-Munksjo Standard Non Wet-Strengthened Qualitative Filter Paper, Grade 617
An extremely fast filtering grade that has a larger particle retention than Grade 631. Grade 617 is a thicker filter paper with higher loading capacity used for rapid filtration of coarse particles and gelatinous precipitates such as in oils and resin solutions.
Supplier: Ahlstrom-Munksjo 61702500
Alert:
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Specifications
35 μm | |
Rapid filtration of coarse particles and gelatinous precipitates such as in oils and resin solutions; construction product testing | |
25 cm |
Fast | |
617 |