missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile CHROMASOLV™ Plus, for HPLC, ≥99.9%, Honeywell Riedel-de Haën™

New Quality CHROMASOLV™ Plus Acetonitrile is suitable for all applications including:Mass spectrometry, HPLC, TLC, GPC, UV, IR, GC, Pesticide Residue Analysis, and RNA/DNA Synthesis and Purification. Product filtered through a 0.2μm filter.

$753.03 - $3,791.63

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecnShow MoreShow Less
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6000231
View Documents
Honeywell-Chromasolv
349981L
1 L N/A
Only null left
 
6000232
View Documents
Honeywell Chemicals
3499825L
2.5 L N/A
Only null left
 
6000233
View Documents
Honeywell Chemicals
349984X25L
4 x 2.5 L N/A
Only null left
 
6000235
View Documents
Honeywell Chemicals
349984X4L
4 x 4 L N/A
Only null left
 
Description

Description

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

P210-P280-P305 + P351 + P338

H225-H302 + H312 + H332-H319

EINECSNumber : 200-835-2

RTECSNumber : AL7700000

SDS
missing translation for 'documents'

missing translation for 'documents'