missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile CHROMASOLV™ Plus, for HPLC, ≥99.9%, Honeywell Riedel-de Haën™
Description
- For HPLC, spectrophotometry, and environmental analysis
- Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
- Low UV impurity profile
- Low impurity gradient LC baseline
- Very low levels of non-volatile impurities
Specifications
Specifications
| Melting Point | -48°C |
| Absorbance | HPLC-gradient/210nm ≤1mAU |
| Density | 0.786g/mL at 25°C |
| Quantity | 4 x 2.5 L |
| Refractive Index | n20/D 1.344 |
| Vapor Pressure | 72.8mmHg (20°C) |
| Linear Formula | CH3CN |
| Beilstein | 741857 |
| Boiling Point | 81°C to 82°C |
| Formula Weight | 41.05g/mol |
| Show More |
Safety and Handling
| Hazard Statement | P210-P280-P305 + P351 + P338 |
| Precautionary Statement | H225-H302 + H312 + H332-H319 |
| EINECSNumber | 200-835-2 |
| RTECSNumber | AL7700000 |