missing translation for 'onlineSavingsMsg'
Learn More

Acetone CHROMASOLV™, for HPLC, ≥99.8%, Honeywell Riedel-de Haën™

Acetone's luminesence intensity is dependent upon the solution components.1 The absorption of UV light by acetone results in its photolysis and the production of radials.1

$207.15 - $1019.48

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetoneShow MoreShow Less
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O
View More Specs

Alert:

Repeated exposure may cause skin dryness or cracking.


Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6000202
View Documents
Honeywell Inc
348501L
1 L
Each for $207.15
Only null left
 
6000203
View Documents
Honeywell Inc
3485025L
2.5 L
Each for $253.83
Only null left
 
6000206
View Documents
Honeywell Inc
348506X1L
6 x 1 L
Case of 6 Each for $1,019.48
Only null left
 
Description

Description

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

P210-P280-P304 + P340 + P312-P305 + P351 + P338-P337 + P313-P403 + P235

H225-H319-H336

EINECSNumber : 200-662-2

RTECSNumber : AL3150000

SDS
missing translation for 'documents'

missing translation for 'documents'