missing translation for 'onlineSavingsMsg'
Learn More

Acetone CHROMASOLV™, for HPLC, ≥99.8%, Solstice

Catalog No. 6000203
Change view
Click to view available options
Quantity:
1 L
2.5 L
6 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000203 2.5 L
6000202 1 L
6000206 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Repeated exposure may cause skin dryness or cracking.
Catalog No. 6000203 Supplier Honeywell Inc Supplier No. 348502.5L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Repeated exposure may cause skin dryness or cracking.

Acetone's luminesence intensity is dependent upon the solution components.1 The absorption of UV light by acetone results in its photolysis and the production of radials.1

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -94°C
Density 0.791g/mL at 25°C
Quantity 2.5 L
Refractive Index n20/D 1.359
Vapor Pressure 184mmHg (20°C)
Linear Formula CH3COCH3
Beilstein 635680
Boiling Point 56°C (760 mmHg)
Formula Weight 58.08g/mol
Percent Purity ≥99.8%
Grade CHROMASOLV
Chemical Name or Material Acetone, for HPLC
Show More Show Less
Hazard Statement P210-P280-P304 + P340 + P312-P305 + P351 + P338-P337 + P313-P403 + P235
Precautionary Statement H225-H319-H336
EINECSNumber 200-662-2
RTECSNumber AL3150000