missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, 99.6%, for analysis

CAS: 64-19-7 | C2H4O2 | 60.05 g/mol

Supplier:  Thermo Scientific Chemicals 295320010

 View more versions of this product

Catalog No. AC295320010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Greener Choice Claims
  • Environmental benefits include:
    • Ozone-Safe or Safe for the Ozone Layer
    • Renewable Energy
    • Free-of a substance that causes environmental harm
    • Non-Toxic
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS P Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only