missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, 99.6%, for analysis
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 16.0°C to 16.5°C |
| Boiling Point | 117.0°C to 118.0°C |
| CAS Min % | 99.5 |
| CAS Max % | 100.0 |
| Assay Percent Range | 99.6% |
| Linear Formula | CH3COOH |
| Beilstein | 02, 96 |
| Fieser | 02,5; 05,3; 07,1; 08,1; 12,568; 13,45; 15,12 |
| Merck Index | 15, 54 |
| Solubility Information | Solubility in water: miscible. Other solubilities: miscible with alcohol, glycerol, ether and, carbon tetrachloride, practically insoluble in carbon disulfide |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS P Statement Causes severe skin burns and eye damage. Flammable liquid and vapor. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. |
RUO – Research Use Only