missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ 6 ft. Logic™ Class II, Type B2 Biosafety Cabinet, US Model
Total exhaust cabinets that provide personnel, product and environmental protection from hazardous particulates requiring BSL 1-4 containment. These models require a dedicated exhaust system and remote blower. All models are NSF/ANSI 49 listed and maximize safety, performance, and comfort.
$43230.12 - $46405.17
Specifications
Description | 6 ft. Logic Class II, Type B2 Biosafety Cabinet |
---|---|
Certifications/Compliance | NSF 49, ETL, UL, CAN/CSA, ADA, ISO 5 Conditions per ISO 14644-1 and 2 |
Chamber Material | Stainless Steel (304) |
Width (Metric) Interior | 184.2 cm |
Product Type | Class II, Type B2 Bio Safety Cabinet |
Catalog Number | Mfr. No. | Plug Type | Includes | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Plug Type | Includes | Price | Quantity | ||||
01-812-553
|
Labconco™
303680002 |
North America (115 V, 15 A) | Cabinet |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
01-812-551
|
Labconco™
303680102 |
North America (115 V, 15 A) | Cabinet, Base Stand |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
01-812-549
|
Labconco™
303681002 |
North America (115 V, 15 A) | Cabinet, Accessory Package |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
01-812-547
|
Labconco™
303681102 |
North America (115 V, 15 A) | Cabinet, Accessory Package, Base Stand |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Performance Features:
- Safe Class II airflows (100% exhaust, remote blower required) that provide personnel, product, and environmental protection.
- Cabinet interior provides ISO Class 5 (Grade A) or better working conditions.
- Constant Airflow Profile™ (CAP) and BalanceFlow™ technology maintain safe downflow – even as HEPA filters load.
- Optimized airflow minimizes energy use and noise output to industry-leading levels.
- Logic OS with 5 in. line-of-sight touchscreen display. Access cabinet information such as filter life, alerts and alarms, data logs.
- Remote connectivity, particle counter integration, and certification records elevate user control of the cabinet
- Smart-Start™ and Night-Smart™ modes simplify startup and shut-down procedures to help ensure consistency and cleanliness before, during, and after work.
- Inclination Technology™ improves ergonomic working conditions by maximizing comfort and accessibility during long periods of work.
- Remote Communication Board for direct communication with remote blower and/or building management system featuring dry contact relays, USB data output and RS-232 RJ45 data output.
- Mag-Zone™ area and mating accessories keep common items such as pipettes and biohazard waste organized and within reach.
- Full, six-year warranty on all Class II biosafety cabinets.
Optional Configurations:
- Accessory Package: UV lamp (254 nm), right-side service fixture with ball-type valve and right and left-side Vacu-Pass™ Cord and Cable Portals
- Telescoping base stand with shelf and leveling feet
Specifications
6 ft. Logic Class II, Type B2 Biosafety Cabinet | |
Stainless Steel (304) | |
Class II, Type B2 Bio Safety Cabinet | |
67 dBA | |
360 W | |
20 cm | |
2348 m³/hr. | |
99.99% | |
199.1 cm | |
8 in. | |
6 Years | |
25.7 in. | |
27 in. | |
57.2 cm | |
32 in. | |
360 W |
NSF 49, ETL, UL, CAN/CSA, ADA, ISO 5 Conditions per ISO 14644-1 and 2 | |
184.2 cm | |
Logic B2 | |
2 Duplex | |
2 Double, Left and Right Side | |
78.4 in. | |
5 in. touchscreen | |
HEPA | |
184.7 cm | |
1382 CFM | |
72.5 in. | |
22.5 in. | |
65.3 cm | |
69 cm | |
81.3 cm |