Learn More
5,5'-Dimethyl-2,2'-bipyridine, 98%
CAS: 1762-34-1 | C12H12N2 | 184.242 g/mol
Supplier: Thermo Scientific Chemicals H2637806
Description
5,5'-Dimethyl-2,2'-bipyridine is widely used as an intermediate in organic synthesis and pharmaceuticals. It is also used as a raw material in the synthesis of dyestuffs and agrochemicals. It is used as a precursor for the preparation of 5-methyl-5âvinyl-2,2â-bipyridine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5,5′-Dimethyl-2,2′-bipyridine is widely used as an intermediate in organic synthesis and pharmaceuticals. It is also used as a raw material in the synthesis of dyestuffs and agrochemicals. It is used as a precursor for the preparation of 5-methyl-5âvinyl-2,2â-bipyridine.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
5,5'-Dimethyl-2,2'-bipyridine | |
114°C to 117°C | |
C12H12N2 | |
5 g | |
PTRATZCAGVBFIQ-UHFFFAOYSA-N | |
5-methyl-2-(5-methylpyridin-2-yl)pyridine | |
15664 | |
98% |
1762-34-1 | |
140°C (3 mmHg) | |
MFCD01740554 | |
5,5'-dimethyl-2,2'-bipyridine, abametapir, 5,5'-dimethyl-2,2'-dipyridyl, 6,6'-bi-3-picoline, 5,5'-dimethyl-2,2'-bipyridyl, 6,6'-di-3-picolyl, 6,6'-di-3-picoline, unii-6uo390amfb, 2,2'-bipyridine, 5,5'-dimethyl, 5,5-dimethyl-2,2-dipyridyl | |
CC1=CN=C(C=C1)C2=NC=C(C=C2)C | |
184.242 | |
184.24 |
Chemical Identifiers
1762-34-1 | |
184.242 | |
PTRATZCAGVBFIQ-UHFFFAOYSA-N | |
15664 | |
CC1=CN=C(C=C1)C2=NC=C(C=C2)C |
C12H12N2 | |
MFCD01740554 | |
5,5'-dimethyl-2,2'-bipyridine, abametapir, 5,5'-dimethyl-2,2'-dipyridyl, 6,6'-bi-3-picoline, 5,5'-dimethyl-2,2'-bipyridyl, 6,6'-di-3-picolyl, 6,6'-di-3-picoline, unii-6uo390amfb, 2,2'-bipyridine, 5,5'-dimethyl, 5,5-dimethyl-2,2-dipyridylShow More | |
5-methyl-2-(5-methylpyridin-2-yl)pyridine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only