missing translation for 'onlineSavingsMsg'
Learn More

5,5'-Dimethyl-2,2'-bipyridine, 98%

CAS: 1762-34-1 | C12H12N2 | 184.242 g/mol

$144.40 - $455.45

Chemical Identifiers

CAS 1762-34-1
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.242
MDL Number MFCD01740554
InChI Key PTRATZCAGVBFIQ-UHFFFAOYSA-N
Synonym 5,5'-dimethyl-2,2'-bipyridine, abametapir, 5,5'-dimethyl-2,2'-dipyridyl, 6,6'-bi-3-picoline, 5,5'-dimethyl-2,2'-bipyridyl, 6,6'-di-3-picolyl, 6,6'-di-3-picoline, unii-6uo390amfb, 2,2'-bipyridine, 5,5'-dimethyl, 5,5-dimethyl-2,2-dipyridyl
PubChem CID 15664
IUPAC Name 5-methyl-2-(5-methylpyridin-2-yl)pyridine
SMILES CC1=CN=C(C=C1)C2=NC=C(C=C2)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2637806
View Documents
Thermo Scientific Chemicals
H2637806
5 g
Each for $144.40
Only null left
 
AAH2637814
View Documents
Thermo Scientific Chemicals
H2637814
25 g
Each for $455.45
Only null left
 
Description

Description

5,5'-Dimethyl-2,2'-bipyridine is widely used as an intermediate in organic synthesis and pharmaceuticals. It is also used as a raw material in the synthesis of dyestuffs and agrochemicals. It is used as a precursor for the preparation of 5-methyl-5’vinyl-2,2’-bipyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,5′-Dimethyl-2,2′-bipyridine is widely used as an intermediate in organic synthesis and pharmaceuticals. It is also used as a raw material in the synthesis of dyestuffs and agrochemicals. It is used as a precursor for the preparation of 5-methyl-5’vinyl-2,2’-bipyridine.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

1762-34-1
184.242
PTRATZCAGVBFIQ-UHFFFAOYSA-N
15664
CC1=CN=C(C=C1)C2=NC=C(C=C2)C
C12H12N2
MFCD01740554
5,5'-dimethyl-2,2'-bipyridine, abametapir, 5,5'-dimethyl-2,2'-dipyridyl, 6,6'-bi-3-picoline, 5,5'-dimethyl-2,2'-bipyridyl, 6,6'-di-3-picolyl, 6,6'-di-3-picoline, unii-6uo390amfb, 2,2'-bipyridine, 5,5'-dimethyl, 5,5-dimethyl-2,2-dipyridyl
5-methyl-2-(5-methylpyridin-2-yl)pyridine

Specifications

1762-34-1
140°C (3 mmHg)
MFCD01740554
5,5'-dimethyl-2,2'-bipyridine, abametapir, 5,5'-dimethyl-2,2'-dipyridyl, 6,6'-bi-3-picoline, 5,5'-dimethyl-2,2'-bipyridyl, 6,6'-di-3-picolyl, 6,6'-di-3-picoline, unii-6uo390amfb, 2,2'-bipyridine, 5,5'-dimethyl, 5,5-dimethyl-2,2-dipyridyl
CC1=CN=C(C=C1)C2=NC=C(C=C2)C
184.242
184.24
5,5'-Dimethyl-2,2'-bipyridine
114°C to 117°C
C12H12N2
5 g
PTRATZCAGVBFIQ-UHFFFAOYSA-N
5-methyl-2-(5-methylpyridin-2-yl)pyridine
15664
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only