missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(4-Iodophenyl)isoxazole, 97%, Thermo Scientific™

Chemical Identifiers
CAS | 160377-48-0 |
---|---|
Molecular Formula | C9H6INO |
Molecular Weight (g/mol) | 271.057 |
MDL Number | MFCD02681898 |
InChI Key | TYNWYXFACGRXMG-UHFFFAOYSA-N |
Synonym | 5-4-iodophenyl isoxazole, 5-4-iodophenyl-1,2-oxazole, isoxazole,5-4-iodophenyl |
PubChem CID | 2776370 |
IUPAC Name | 5-(4-iodophenyl)-1,2-oxazole |
SMILES | C1=CC(=CC=C1C2=CC=NO2)I |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
CC12911CB
|
Thermo Scientific Chemicals
CC12911CB |
250mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC12911DA
|
Thermo Scientific Chemicals
CC12911DA |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC12911DE
|
Thermo Scientific Chemicals
CC12911DE |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
160377-48-0 | |
271.057 | |
TYNWYXFACGRXMG-UHFFFAOYSA-N | |
2776370 | |
C1=CC(=CC=C1C2=CC=NO2)I |
C9H6INO | |
MFCD02681898 | |
5-4-iodophenyl isoxazole, 5-4-iodophenyl-1,2-oxazole, isoxazole,5-4-iodophenylShow More | |
5-(4-iodophenyl)-1,2-oxazole |
Specifications
160377-48-0 | |
100.0 | |
Amber glass bottle | |
MFCD02681898 | |
5-4-iodophenyl isoxazole, 5-4-iodophenyl-1,2-oxazole, isoxazole,5-4-iodophenyl | |
C1=CC(=CC=C1C2=CC=NO2)I | |
271.057 | |
271.05 | |
5-(4-Iodophenyl)isoxazole |
96.5 | |
97% | |
C9H6INO | |
250mg | |
TYNWYXFACGRXMG-UHFFFAOYSA-N | |
5-(4-iodophenyl)-1,2-oxazole | |
2776370 | |
97% |