missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(4-Iodophenyl)isoxazole, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC12911CB
Specifications
| 5-(4-Iodophenyl)isoxazole | |
| 96.5 | |
| 97% | |
| C9H6INO | |
| 250mg | |
| TYNWYXFACGRXMG-UHFFFAOYSA-N | |
| 5-(4-iodophenyl)-1,2-oxazole | |
| 2776370 | |
| 97% |
| 160377-48-0 | |
| 100.0 | |
| Amber glass bottle | |
| MFCD02681898 | |
| 5-4-iodophenyl isoxazole, 5-4-iodophenyl-1,2-oxazole, isoxazole,5-4-iodophenyl | |
| C1=CC(=CC=C1C2=CC=NO2)I | |
| 271.057 | |
| 271.05 |
Chemical Identifiers
| 160377-48-0 | |
| 271.057 | |
| TYNWYXFACGRXMG-UHFFFAOYSA-N | |
| 2776370 | |
| C1=CC(=CC=C1C2=CC=NO2)I |
| C9H6INO | |
| MFCD02681898 | |
| 5-4-iodophenyl isoxazole, 5-4-iodophenyl-1,2-oxazole, isoxazole,5-4-iodophenyl | |
| 5-(4-iodophenyl)-1,2-oxazole |