Learn More
4'-Hydroxybiphenyl-4-carboxylic acid, 99%
CAS: 58574-03-1 | C13H10O3 | 214.22 g/mol
Supplier: Thermo Scientific Chemicals B2194688
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4'-Hydroxybiphenyl-4-carboxylic acid | |
292°C to 295°C | |
MFCD00059078 | |
4'-hydroxy-4-biphenylcarboxylic acid, 4-4-hydroxyphenyl benzoic acid, 4'-hydroxybiphenyl-4-carboxylic acid, 4'-hydroxy-1,1'-biphenyl-4-carboxylic acid, 4-hydroxy-4'-biphenylcarboxylic acid, 4'-hydroxy biphenyl-4-carboxylic acid, 4-hydroxybiphenyl-4'-carboxylic acid, 1,1'-biphenyl-4-carboxylic acid, 4'-hydroxy, chembl90522, 4'-hydroxy 1,1'-biphenyl-4-carboxylic acid | |
C1=CC(=CC=C1C2=CC=C(C=C2)O)C(=O)O | |
214.22 | |
214.22 |
58574-03-1 | |
C13H10O3 | |
2.5 g | |
JTGCXYYDAVPSFD-UHFFFAOYSA-N | |
4-(4-hydroxyphenyl)benzoic acid | |
301556 | |
99% |
Chemical Identifiers
58574-03-1 | |
214.22 | |
JTGCXYYDAVPSFD-UHFFFAOYSA-N | |
301556 | |
C1=CC(=CC=C1C2=CC=C(C=C2)O)C(=O)O |
C13H10O3 | |
MFCD00059078 | |
4'-hydroxy-4-biphenylcarboxylic acid, 4-4-hydroxyphenyl benzoic acid, 4'-hydroxybiphenyl-4-carboxylic acid, 4'-hydroxy-1,1'-biphenyl-4-carboxylic acid, 4-hydroxy-4'-biphenylcarboxylic acid, 4'-hydroxy biphenyl-4-carboxylic acid, 4-hydroxybiphenyl-4'-carboxylic acid, 1,1'-biphenyl-4-carboxylic acid, 4'-hydroxy, chembl90522, 4'-hydroxy 1,1'-biphenyl-4-carboxylic acidShow More | |
4-(4-hydroxyphenyl)benzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only