Learn More
4-Hydrazino-2,6-dimethylpyrimidine, ≥95%, Thermo Scientific™

Chemical Identifiers
CAS | 14331-56-7 |
---|---|
Molecular Formula | C6H10N4 |
Molecular Weight (g/mol) | 138.174 |
MDL Number | MFCD00102150 |
InChI Key | HVAYIAWJHRUOBC-UHFFFAOYSA-N |
Synonym | 4-hydrazino-2,6-dimethylpyrimidine, 4-hydrazinyl-2,6-dimethylpyrimidine, 2,6-dimethyl-4-hydrazinopyrimidine, 2,6-dimethylpyrimidin-4-yl hydrazine, 2,6-dimethyl-pyrimidin-4-yl-hydrazine, 2,4-dimethyl-6-hydra-zinopyrimidine, 2,6-dimethylpyrimidine-4-ylhydrazine, 2,6-dimethyl-4-pyrimidinyl hydrazine, pyrimidine,4-hydrazinyl-2,6-dimethyl, pyrimidine, 4-hydrazinyl-2,6-dimethylShow More |
PubChem CID | 736395 |
IUPAC Name | (2,6-dimethylpyrimidin-4-yl)hydrazine |
SMILES | CC1=CC(=NC(=N1)C)NN |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
BTB10371CB
|
Thermo Scientific Chemicals
BTB10371CB |
250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTB10371DA
|
Thermo Scientific Chemicals
BTB10371DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
14331-56-7 | |
138.174 | |
HVAYIAWJHRUOBC-UHFFFAOYSA-N | |
736395 | |
CC1=CC(=NC(=N1)C)NN |
C6H10N4 | |
MFCD00102150 | |
4-hydrazino-2,6-dimethylpyrimidine, 4-hydrazinyl-2,6-dimethylpyrimidine, 2,6-dimethyl-4-hydrazinopyrimidine, 2,6-dimethylpyrimidin-4-yl hydrazine, 2,6-dimethyl-pyrimidin-4-yl-hydrazine, 2,4-dimethyl-6-hydra-zinopyrimidine, 2,6-dimethylpyrimidine-4-ylhydrazine, 2,6-dimethyl-4-pyrimidinyl hydrazine, pyrimidine,4-hydrazinyl-2,6-dimethyl, pyrimidine, 4-hydrazinyl-2,6-dimethylShow More | |
(2,6-dimethylpyrimidin-4-yl)hydrazine |
Specifications
14331-56-7 | |
100.0 | |
Amber Glass Bottle | |
MFCD00102150 | |
4-hydrazino-2,6-dimethylpyrimidine, 4-hydrazinyl-2,6-dimethylpyrimidine, 2,6-dimethyl-4-hydrazinopyrimidine, 2,6-dimethylpyrimidin-4-yl hydrazine, 2,6-dimethyl-pyrimidin-4-yl-hydrazine, 2,4-dimethyl-6-hydra-zinopyrimidine, 2,6-dimethylpyrimidine-4-ylhydrazine, 2,6-dimethyl-4-pyrimidinyl hydrazine, pyrimidine,4-hydrazinyl-2,6-dimethyl, pyrimidine, 4-hydrazinyl-2,6-dimethyl | |
CC1=CC(=NC(=N1)C)NN | |
138.174 | |
138.17 | |
4-Hydrazino-2, 6-dimethylpyrimidine |
94.5 | |
95% | |
C6H10N4 | |
250mg | |
HVAYIAWJHRUOBC-UHFFFAOYSA-N | |
(2,6-dimethylpyrimidin-4-yl)hydrazine | |
736395 | |
95% |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning