Learn More
4-Hydrazino-2,6-dimethylpyrimidine, ≥95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals BTB10371CB
Specifications
| 4-Hydrazino-2, 6-dimethylpyrimidine | |
| 94.5 | |
| 95% | |
| C6H10N4 | |
| 250mg | |
| HVAYIAWJHRUOBC-UHFFFAOYSA-N | |
| (2,6-dimethylpyrimidin-4-yl)hydrazine | |
| 736395 | |
| 95% |
| 14331-56-7 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD00102150 | |
| 4-hydrazino-2,6-dimethylpyrimidine, 4-hydrazinyl-2,6-dimethylpyrimidine, 2,6-dimethyl-4-hydrazinopyrimidine, 2,6-dimethylpyrimidin-4-yl hydrazine, 2,6-dimethyl-pyrimidin-4-yl-hydrazine, 2,4-dimethyl-6-hydra-zinopyrimidine, 2,6-dimethylpyrimidine-4-ylhydrazine, 2,6-dimethyl-4-pyrimidinyl hydrazine, pyrimidine,4-hydrazinyl-2,6-dimethyl, pyrimidine, 4-hydrazinyl-2,6-dimethyl | |
| CC1=CC(=NC(=N1)C)NN | |
| 138.174 | |
| 138.17 |
Chemical Identifiers
| 14331-56-7 | |
| 138.174 | |
| HVAYIAWJHRUOBC-UHFFFAOYSA-N | |
| 736395 | |
| CC1=CC(=NC(=N1)C)NN |
| C6H10N4 | |
| MFCD00102150 | |
| 4-hydrazino-2,6-dimethylpyrimidine, 4-hydrazinyl-2,6-dimethylpyrimidine, 2,6-dimethyl-4-hydrazinopyrimidine, 2,6-dimethylpyrimidin-4-yl hydrazine, 2,6-dimethyl-pyrimidin-4-yl-hydrazine, 2,4-dimethyl-6-hydra-zinopyrimidine, 2,6-dimethylpyrimidine-4-ylhydrazine, 2,6-dimethyl-4-pyrimidinyl hydrazine, pyrimidine,4-hydrazinyl-2,6-dimethyl, pyrimidine, 4-hydrazinyl-2,6-dimethyl | |
| (2,6-dimethylpyrimidin-4-yl)hydrazine |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning