missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-nitrophenol, 98%, Thermo Scientificâ„¢

Chemical Identifiers

CAS 7693-52-9
Molecular Formula C6H4BrNO3
Molecular Weight (g/mol) 218.01
MDL Number MFCD00082540
InChI Key CUTFAPGINUFNQM-UHFFFAOYSA-N
Synonym 2-nitro-4-bromophenol, phenol, 4-bromo-2-nitro, 4-bromo-2-nitro-phenol, 4-bromo-1-hydroxy-2-nitrobenzene, phenol, 2-nitro-5-bromo, pubchem4099, 4-brom-2-nitro-phenol, acmc-209p6y, 4-06-00-01363 beilstein handbook reference, ksc494e2jShow MoreShow Less
PubChem CID 24364
IUPAC Name 4-bromo-2-nitrophenol
SMILES OC1=CC=C(Br)C=C1[N+]([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC367920050
SDS View Documents
Thermo Scientific Chemicals
367920050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
W

GHS Signal Word: Warning

EINECSNumber : 231-707-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only