Learn More
4-Bromo-2-fluorobenzoyl chloride, 99%

CAS: 151982-51-3 | C7H3BrClFO | 237.46 g/mol
$86.57 - $287.75
Chemical Identifiers
CAS | 151982-51-3 |
---|---|
Molecular Formula | C7H3BrClFO |
Molecular Weight (g/mol) | 237.46 |
MDL Number | MFCD03411584 |
InChI Key | PCFIABOQFAFDAU-UHFFFAOYSA-N |
Synonym | 2-fluoro-4-bromobenzoyl chloride, benzoyl chloride, 4-bromo-2-fluoro, benzoyl chloride,4-bromo-2-fluoro, acmc-20amnm, 4-bromo-2-fluorobenzoylchloride, 4-bromo-2-fluoro-benzoyl chloride, 4-bromo-2-fluorobenzoic acid chloride, 4-bromanyl-2-fluoranyl-benzoyl chlorideShow More |
PubChem CID | 2734026 |
IUPAC Name | 4-bromo-2-fluorobenzoyl chloride |
SMILES | C1=CC(=C(C=C1Br)F)C(=O)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC347460050
|
Thermo Scientific Chemicals
347460050 |
5 g | Glass bottle |
Each for $86.57
|
|
|||||
AC347460250
|
Thermo Scientific Chemicals
347460250 |
25 g | Glass bottle |
Each for $287.75
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
151982-51-3 | |
237.46 | |
PCFIABOQFAFDAU-UHFFFAOYSA-N | |
2734026 | |
C1=CC(=C(C=C1Br)F)C(=O)Cl |
C7H3BrClFO | |
MFCD03411584 | |
2-fluoro-4-bromobenzoyl chloride, benzoyl chloride, 4-bromo-2-fluoro, benzoyl chloride,4-bromo-2-fluoro, acmc-20amnm, 4-bromo-2-fluorobenzoylchloride, 4-bromo-2-fluoro-benzoyl chloride, 4-bromo-2-fluorobenzoic acid chloride, 4-bromanyl-2-fluoranyl-benzoyl chlorideShow More | |
4-bromo-2-fluorobenzoyl chloride |
Specifications
151982-51-3 | |
100.0 | |
White to Yellow | |
Authentic | |
Glass bottle | |
MFCD03411584 | |
2-fluoro-4-bromobenzoyl chloride, benzoyl chloride, 4-bromo-2-fluoro, benzoyl chloride,4-bromo-2-fluoro, acmc-20amnm, 4-bromo-2-fluorobenzoylchloride, 4-bromo-2-fluoro-benzoyl chloride, 4-bromo-2-fluorobenzoic acid chloride, 4-bromanyl-2-fluoranyl-benzoyl chloride | |
PCFIABOQFAFDAU-UHFFFAOYSA-N | |
4-bromo-2-fluorobenzoyl chloride | |
2734026 | |
99% | |
4-Bromo-2-fluorobenzoyl chloride, 99% |
98.5 | |
24.0°C to 24.5°C | |
86.0°C to 88.0°C (15.0 mmHg) | |
98.5% min. (GC) | |
C7H3BrClFO | |
5 g | |
Solubility in water: hydrdolyses | |
C1=CC(=C(C=C1Br)F)C(=O)Cl | |
237.46 | |
237.46 | |
Crystalline Mass or Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
RUO – Research Use Only