Learn More
4-[2-(6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinolinyl)ethyl]aniline, 97%
CAS: 82925-02-8 | C19H24N2O2 | 312.41 g/mol
Supplier: Thermo Scientific Chemicals H5415606
Description
It is an intermediate for the preparation of Elacridar.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an intermediate for the preparation of Elacridar.
Solubility
Soluble in dichloromethane, ethyl acetate and methanol.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications
4-[2-(6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinolinyl)ethyl]aniline | |
C19H24N2O2 | |
5 g | |
Soluble in dichloromethane,ethyl acetate and methanol. | |
COC1=C(OC)C=C2CN(CCC3=CC=C(N)C=C3)CCC2=C1 | |
312.41 | |
312.41 |
82925-02-8 | |
MFCD04974539 | |
4-2-6,7-dimethoxy-3,4-dihydroisoquinolin-2 1h-yl ethyl aniline, 4-2-6,7-dimethoxy-1,2,3,4-tetrahydro-2-isoquinolinyl ethyl aniline, 4-2-3,4-dihydro-6,7-dimethoxy-2 1h-isoquinolinyl ethyl-benzenamine, 4-2-6,7-dimethoxy-3,4-dihydro-1h-isoquinolin-2-yl ethyl aniline, benzenamine,4-2-3,4-dihydro-6,7-dimethoxy-2 1h-isoquinolinyl ethyl, 4-2-6,7-dimethoxy-3,4-dihydroisoquinoline-2 1h-yl-ethyl-benzenamine, 4-2-1,2,3,4-tetrahydro-6,7-dimethoxy-2-isoquinolyl ethyl benzenamine, 4-2-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline-2-yl ethyl benzenamine, 4-2-6,7-dimethoxy-3,4-dihydroisoquinolin-2 1h-yl ethyl benzenamine | |
DGOOLMGPMIHRFY-UHFFFAOYSA-N | |
4-[2-(6,7-dimethoxy-3,4-dihydro-1H-isoquinolin-2-yl)ethyl]aniline | |
10041227 | |
97% |
Chemical Identifiers
82925-02-8 | |
312.41 | |
DGOOLMGPMIHRFY-UHFFFAOYSA-N | |
10041227 | |
COC1=C(OC)C=C2CN(CCC3=CC=C(N)C=C3)CCC2=C1 |
C19H24N2O2 | |
MFCD04974539 | |
4-2-6,7-dimethoxy-3,4-dihydroisoquinolin-2 1h-yl ethyl aniline, 4-2-6,7-dimethoxy-1,2,3,4-tetrahydro-2-isoquinolinyl ethyl aniline, 4-2-3,4-dihydro-6,7-dimethoxy-2 1h-isoquinolinyl ethyl-benzenamine, 4-2-6,7-dimethoxy-3,4-dihydro-1h-isoquinolin-2-yl ethyl aniline, benzenamine,4-2-3,4-dihydro-6,7-dimethoxy-2 1h-isoquinolinyl ethyl, 4-2-6,7-dimethoxy-3,4-dihydroisoquinoline-2 1h-yl-ethyl-benzenamine, 4-2-1,2,3,4-tetrahydro-6,7-dimethoxy-2-isoquinolyl ethyl benzenamine, 4-2-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline-2-yl ethyl benzenamine, 4-2-6,7-dimethoxy-3,4-dihydroisoquinolin-2 1h-yl ethyl benzenamineShow More | |
4-[2-(6,7-dimethoxy-3,4-dihydro-1H-isoquinolin-2-yl)ethyl]aniline |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only