missing translation for 'onlineSavingsMsg'
Learn More

4-[2-(6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinolinyl)ethyl]aniline, 97%

Catalog No. p-7035926
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5415603 1 g
AAH5415606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5415603 Supplier Thermo Scientific Chemicals Supplier No. H5415603
Only null left

CAS: 82925-02-8 | C19H24N2O2 | 312.41 g/mol

It is an intermediate for the preparation of Elacridar.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate for the preparation of Elacridar.

Solubility
Soluble in dichloromethane, ethyl acetate and methanol.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 82925-02-8
Molecular Formula C19H24N2O2
Molecular Weight (g/mol) 312.41
MDL Number MFCD04974539
InChI Key DGOOLMGPMIHRFY-UHFFFAOYSA-N
Synonym 4-2-6,7-dimethoxy-3,4-dihydroisoquinolin-2 1h-yl ethyl aniline, 4-2-6,7-dimethoxy-1,2,3,4-tetrahydro-2-isoquinolinyl ethyl aniline, 4-2-3,4-dihydro-6,7-dimethoxy-2 1h-isoquinolinyl ethyl-benzenamine, 4-2-6,7-dimethoxy-3,4-dihydro-1h-isoquinolin-2-yl ethyl aniline, benzenamine,4-2-3,4-dihydro-6,7-dimethoxy-2 1h-isoquinolinyl ethyl, 4-2-6,7-dimethoxy-3,4-dihydroisoquinoline-2 1h-yl-ethyl-benzenamine, 4-2-1,2,3,4-tetrahydro-6,7-dimethoxy-2-isoquinolyl ethyl benzenamine, 4-2-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline-2-yl ethyl benzenamine, 4-2-6,7-dimethoxy-3,4-dihydroisoquinolin-2 1h-yl ethyl benzenamine
PubChem CID 10041227
IUPAC Name 4-[2-(6,7-dimethoxy-3,4-dihydro-1H-isoquinolin-2-yl)ethyl]aniline
SMILES COC1=C(OC)C=C2CN(CCC3=CC=C(N)C=C3)CCC2=C1

Specifications

Quantity 1 g
Solubility Information Soluble in dichloromethane,ethyl acetate and methanol.
Formula Weight 312.41
Percent Purity 97%
Chemical Name or Material 4-[2-(6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinolinyl)ethyl]aniline
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only