missing translation for 'onlineSavingsMsg'
Learn More

3M™ Peltor™ PTL™ Ear Muffs

Allow workers to listen safely—no need for removal

$83.54 - $83.54

Specifications

Color Black
Certifications/Compliance UL
Cushion Material Liquid-foam
For Use With (Application) Assembly, cleaning, demolition, facility maintenance, grinding, machine operations, painting, sanding, welding
Product Type Ear Muffs
View More Specs

Alert:

The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.


Products 1
Catalog Number Mfr. No. Style Price Quantity  
Catalog Number Mfr. No. Style Price Quantity  
19-130-1976
View Documents
3M™
H7P3EPTL
Cap-attached
Each for $83.54
Pack of 10 Each for $786.90
Only null left
 
Description

Description

A simple touch of a button activates the Push-to-Listen (PTL)† feature for a 30-second interval. This allows the user to hear speech and sound outside the muff cup, clearly and safely amplified up to 87dB.

  • Pivoting earcups tilt easily to fit most facial profiles
  • Liquid foam-filled ear cushions for comfortable fit
  • NRR: 26dB (over-the-head); 25dB (cap attached)
  • 3M Catalog# H7A-PTL , 3M Catalog# H7P3E-PTL

Over-the-Head Model features padded headband with four-point suspension
Cap-Mounted Model fits most standard slotted hardhats

† The Push-to-Listen feature is in one earcup only; other cup is standard protective style.

Specifications

Specifications

Black
Liquid-foam
Ear Muffs
Foam, Plastic, Metal
UL
Assembly, cleaning, demolition, facility maintenance, grinding, machine operations, painting, sanding, welding
No
Padded headband
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'