3M™ Peltor™ PTL™ Ear Muffs
Allow workers to listen safely—no need for removal
Supplier:  3M™ H7P3EPTL
 View more versions of this product
Catalog No. 19-130-1976
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Description
Description
A simple touch of a button activates the Push-to-Listen (PTL)† feature for a 30-second interval. This allows the user to hear speech and sound outside the muff cup, clearly and safely amplified up to 87dB.
  • Pivoting earcups tilt easily to fit most facial profiles
  • Liquid foam-filled ear cushions for comfortable fit
  • NRR: 26dB (over-the-head); 25dB (cap attached)
  • 3M Catalog# H7A-PTL , 3M Catalog# H7P3E-PTL
Over-the-Head Model features padded headband with four-point suspension
Cap-Mounted Model fits most standard slotted hardhats
† The Push-to-Listen feature is in one earcup only; other cup is standard protective style.
Specifications
Specifications
Padded headband
UL
Assembly, cleaning, demolition, facility maintenance, grinding, machine operations, painting, sanding, welding
25 dB
No
Cap-attached
Black
Liquid-foam
H7P3E-PTL
Ear Muffs
Foam, Plastic, Metal