Learn More
3-Methoxy-1-butanol, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 125790010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Environmental benefits include:
- Energy Star Certification
- Source Reduction
Specifications
3-Methoxy-1-butanol | |
2517-43-3 | |
100.0 | |
161.0°C | |
Authentic | |
Glass bottle | |
1.4150 to 1.4165 | |
MFCD00002931 | |
1987 | |
3-methoxy-1-butanol, 3-methoxybutanol, 1-butanol, 3-methoxy, methoxybutanol, ccris 8976, 1-butanol, 3-methoxy-,-, 1-butanol, 3-methoxy-, r, 3-methoxy-butanol, 3-methoxybutane-1-ol, 3-methoxybutyl alcohol | |
JSGVZVOGOQILFM-UHFFFAOYNA-N | |
3-methoxybutan-1-ol | |
17291 | |
99% |
99% | |
98.5 | |
0.9200g/mL | |
46°C | |
98.5% min. (GC) | |
C5H12O2 | |
CH3CH(OCH3)CH2CH2OH | |
1L | |
0.92 | |
Solubility in water: soluble. Other solubilities: miscible with most common organic solvents | |
COC(C)CCO | |
104.15 | |
104.15 |
Chemical Identifiers
2517-43-3 | |
104.15 | |
JSGVZVOGOQILFM-UHFFFAOYNA-N | |
17291 | |
COC(C)CCO |
C5H12O2 | |
MFCD00002931 | |
3-methoxy-1-butanol, 3-methoxybutanol, 1-butanol, 3-methoxy, methoxybutanol, ccris 8976, 1-butanol, 3-methoxy-,-, 1-butanol, 3-methoxy-, r, 3-methoxy-butanol, 3-methoxybutane-1-ol, 3-methoxybutyl alcoholShow More | |
3-methoxybutan-1-ol |
Safety and Handling
GHS H Statement:
Flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge
GHS Signal Word: Warning
EINECSNumber : 219-741-8
TSCA : TSCA
RUO â Research Use Only