missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-1-butanol, 99%, Thermo Scientific™

Chemical Identifiers

CAS 2517-43-3
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00002931
InChI Key JSGVZVOGOQILFM-UHFFFAOYNA-N
Synonym 3-methoxy-1-butanol, 3-methoxybutanol, 1-butanol, 3-methoxy, methoxybutanol, ccris 8976, 1-butanol, 3-methoxy-,-, 1-butanol, 3-methoxy-, r, 3-methoxy-butanol, 3-methoxybutane-1-ol, 3-methoxybutyl alcohol
PubChem CID 17291
IUPAC Name 3-methoxybutan-1-ol
SMILES COC(C)CCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC125791000
SDS View Documents
Thermo Scientific Chemicals
125791000
100mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC125790010
SDS View Documents
Thermo Scientific Chemicals
125790010
1L Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2517-43-3
104.15
JSGVZVOGOQILFM-UHFFFAOYNA-N
17291
COC(C)CCO
C5H12O2
MFCD00002931
3-methoxy-1-butanol, 3-methoxybutanol, 1-butanol, 3-methoxy, methoxybutanol, ccris 8976, 1-butanol, 3-methoxy-,-, 1-butanol, 3-methoxy-, r, 3-methoxy-butanol, 3-methoxybutane-1-ol, 3-methoxybutyl alcohol
3-methoxybutan-1-ol

Specifications

2517-43-3
100.0
161.0°C
Authentic
Glass bottle
1.4150 to 1.4165
MFCD00002931
1987
3-methoxy-1-butanol, 3-methoxybutanol, 1-butanol, 3-methoxy, methoxybutanol, ccris 8976, 1-butanol, 3-methoxy-,-, 1-butanol, 3-methoxy-, r, 3-methoxy-butanol, 3-methoxybutane-1-ol, 3-methoxybutyl alcohol
JSGVZVOGOQILFM-UHFFFAOYNA-N
3-methoxybutan-1-ol
17291
99%
98.5
0.9200g/mL
46°C
98.5% min. (GC)
C5H12O2
CH3CH(OCH3)CH2CH2OH
100mL
0.92
Solubility in water: soluble. Other solubilities: miscible with most common organic solvents
COC(C)CCO
104.15
104.15
3-Methoxy-1-butanol, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge

GHS Signal Word: Warning

EINECSNumber : 219-741-8

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only