Learn More
2-Methyl-2-pentenoic acid, 99%
CAS: 3142-72-1 | C6H10O2 | 114.144 g/mol
$59.42 - $835.77
Chemical Identifiers
CAS | 3142-72-1 |
---|---|
Molecular Formula | C6H10O2 |
Molecular Weight (g/mol) | 114.144 |
MDL Number | MFCD00002655 |
InChI Key | JJYWRQLLQAKNAD-SNAWJCMRSA-N |
Synonym | 2-methyl-2-pentenoic acid, trans-2-methyl-2-pentenoic acid, e-2-methylpent-2-enoic acid, e-2-methyl-2-pentenoic acid, 2-pentenoic acid, 2-methyl, 2-pentenoic acid, 2-methyl-, e, e-2-methylpent-2-en-1-oic acid, s-2-methylpentenoic acid, 2e-2-methylpent-2-enoic acid, 2-methylpent-2-en-1-oic acidShow More |
PubChem CID | 5365909 |
IUPAC Name | (E)-2-methylpent-2-enoic acid |
SMILES | CCC=C(C)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1658709
|
Thermo Scientific Chemicals
A1658709 |
10 g |
Each for $59.42
|
|
||||||
AAA1658718
|
Thermo Scientific Chemicals
A1658718 |
50 g |
Each for $216.55
|
|
||||||
AAA1658730
|
Thermo Scientific Chemicals
A1658730 |
250 g |
Each for $835.77
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3142-72-1 | |
114.144 | |
JJYWRQLLQAKNAD-SNAWJCMRSA-N | |
5365909 | |
CCC=C(C)C(=O)O |
C6H10O2 | |
MFCD00002655 | |
2-methyl-2-pentenoic acid, trans-2-methyl-2-pentenoic acid, e-2-methylpent-2-enoic acid, e-2-methyl-2-pentenoic acid, 2-pentenoic acid, 2-methyl, 2-pentenoic acid, 2-methyl-, e, e-2-methylpent-2-en-1-oic acid, s-2-methylpentenoic acid, 2e-2-methylpent-2-enoic acid, 2-methylpent-2-en-1-oic acidShow More | |
(E)-2-methylpent-2-enoic acid |
Specifications
3142-72-1 | |
120°C (20 mmHg) | |
Characteristic | |
MFCD00002655 | |
UN3265 | |
JJYWRQLLQAKNAD-SNAWJCMRSA-N | |
(E)-2-methylpent-2-enoic acid | |
5365909 | |
99% |
0.978 | |
107°C (225°F) | |
C6H10O2 | |
10 g | |
2-methyl-2-pentenoic acid, trans-2-methyl-2-pentenoic acid, e-2-methylpent-2-enoic acid, e-2-methyl-2-pentenoic acid, 2-pentenoic acid, 2-methyl, 2-pentenoic acid, 2-methyl-, e, e-2-methylpent-2-en-1-oic acid, s-2-methylpentenoic acid, 2e-2-methylpent-2-enoic acid, 2-methylpent-2-en-1-oic acid | |
CCC=C(C)C(=O)O | |
114.144 | |
114.15 | |
2-Methyl-2-pentenoic acid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 221-552-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only