missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-pentenoic acid, 99%

CAS: 3142-72-1 | C6H10O2 | 114.144 g/mol

Supplier:  Thermo Scientific Chemicals A1658730

 View more versions of this product

Catalog No. AAA1658730


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Methyl-2-pentenoic acid
0.978
107°C (225°F)
C6H10O2
250 g
2-methyl-2-pentenoic acid, trans-2-methyl-2-pentenoic acid, e-2-methylpent-2-enoic acid, e-2-methyl-2-pentenoic acid, 2-pentenoic acid, 2-methyl, 2-pentenoic acid, 2-methyl-, e, e-2-methylpent-2-en-1-oic acid, s-2-methylpentenoic acid, 2e-2-methylpent-2-enoic acid, 2-methylpent-2-en-1-oic acid
CCC=C(C)C(=O)O
114.144
114.15
3142-72-1
120°C (20 mmHg)
Characteristic
MFCD00002655
UN3265
JJYWRQLLQAKNAD-SNAWJCMRSA-N
(E)-2-methylpent-2-enoic acid
5365909
99%

Chemical Identifiers

3142-72-1
114.144
JJYWRQLLQAKNAD-SNAWJCMRSA-N
5365909
CCC=C(C)C(=O)O
C6H10O2
MFCD00002655
2-methyl-2-pentenoic acid, trans-2-methyl-2-pentenoic acid, e-2-methylpent-2-enoic acid, e-2-methyl-2-pentenoic acid, 2-pentenoic acid, 2-methyl, 2-pentenoic acid, 2-methyl-, e, e-2-methylpent-2-en-1-oic acid, s-2-methylpentenoic acid, 2e-2-methylpent-2-enoic acid, 2-methylpent-2-en-1-oic acid
(E)-2-methylpent-2-enoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 221-552-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only