Learn More
2-Ethylhexyl 4-dimethylaminobenzoate, 99%
CAS: 21245-02-3 | C17H27NO2 | 277.408 g/mol
Supplier: Thermo Scientific Chemicals H54136AE
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Environmental benefits include:
- Renewable Energy
Specifications
2-Ethylhexyl 4-dimethylaminobenzoate | |
0.995 | |
113°C (235°F) | |
1.542 | |
100 mL | |
2-ethylhexyl 4-dimethylamino benzoate, padimate o, escalol 507, padimate-o, octyl dimethyl paba, arlatone uvb, benzoic acid, 4-dimethylamino-, 2-ethylhexyl ester, 2-ethylhexyl p-dimethylaminobenzoate, eusolex 6007, sundown | |
CCCCC(CC)COC(=O)C1=CC=C(C=C1)N(C)C | |
277.408 | |
277.4 |
21245-02-3 | |
325°C | |
C17H27NO2 | |
MFCD00017526 | |
14,3232 | |
WYWZRNAHINYAEF-UHFFFAOYSA-N | |
2-ethylhexyl 4-(dimethylamino)benzoate | |
30541 | |
99% |
Chemical Identifiers
21245-02-3 | |
277.408 | |
WYWZRNAHINYAEF-UHFFFAOYSA-N | |
30541 | |
CCCCC(CC)COC(=O)C1=CC=C(C=C1)N(C)C |
C17H27NO2 | |
MFCD00017526 | |
2-ethylhexyl 4-dimethylamino benzoate, padimate o, escalol 507, padimate-o, octyl dimethyl paba, arlatone uvb, benzoic acid, 4-dimethylamino-, 2-ethylhexyl ester, 2-ethylhexyl p-dimethylaminobenzoate, eusolex 6007, sundownShow More | |
2-ethylhexyl 4-(dimethylamino)benzoate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P281-P308+P313-P501c
H360
EINECSNumber : 244-289-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only