missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl 4-dimethylaminobenzoate, 99%

Catalog No. p-7035909
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54136AE 100 mL
AAH54136AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH54136AE Supplier Thermo Scientific Chemicals Supplier No. H54136AE
Only null left

CAS: 21245-02-3 | C17H27NO2 | 277.408 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21245-02-3
Molecular Formula C17H27NO2
Molecular Weight (g/mol) 277.408
MDL Number MFCD00017526
InChI Key WYWZRNAHINYAEF-UHFFFAOYSA-N
Synonym 2-ethylhexyl 4-dimethylamino benzoate, padimate o, escalol 507, padimate-o, octyl dimethyl paba, arlatone uvb, benzoic acid, 4-dimethylamino-, 2-ethylhexyl ester, 2-ethylhexyl p-dimethylaminobenzoate, eusolex 6007, sundown
PubChem CID 30541
IUPAC Name 2-ethylhexyl 4-(dimethylamino)benzoate
SMILES CCCCC(CC)COC(=O)C1=CC=C(C=C1)N(C)C

Specifications

Density 0.995
Boiling Point 325°C
Flash Point 113°C (235°F)
Refractive Index 1.542
Quantity 100 mL
Merck Index 14,3232
Formula Weight 277.4
Percent Purity 99%
Chemical Name or Material 2-Ethylhexyl 4-dimethylaminobenzoate
Hazard Category H360
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 244-289-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only