Learn More
2-Chlorotrityl chloride on polystyrene, 1% cross-linked, 100-200 mesh, 1.0-1.4 mmol/g
CAS: 42074-68-0 | C19H14Cl2 | 313.221 g/mol
Supplier: Thermo Scientific Chemicals 04440704
Description
2-Chlorotrityl chloride is used as an intermediate of clotrimazole. It is also used in convergent peptide synthesis. Furthermore, it is used in the preparation of modified trityl nucleosides. In addition, it serves as a protecting group reagent for carboxylic acids forming esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Environmental benefits include:
- Recyclable
- Recycled Content
- Renewable Materials
Specifications
2-Chlorotrityl chloride on polystyrene | |
Powder | |
C19H14Cl2 | |
2-chlorotrityl chloride, chloro 2-chlorophenyl methylene dibenzene, 1-chloro-2-chlorodiphenylmethyl benzene, 2-chlorophenyldiphenylmethyl chloride, 2-chlorotrityl chloride resin, 2-chlorophenyl diphenylmethyl chloride, 2-chlorotrityl resin, 2-chlorotritylchloride, chloro 2-chlorophenyl diphenylmethane, 2-chlorotrityl chloride, polymer-bound | |
JFLSOKIMYBSASW-UHFFFAOYSA-N | |
1-chloro-2-[chloro(diphenyl)methyl]benzene | |
94524 |
1% cross-linked, 1.0 to 1.4 mmol/g | |
42074-68-0 | |
MFCD00040399 | |
2 g | |
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3Cl)Cl | |
313.221 | |
100 to 200 Mesh |
Chemical Identifiers
42074-68-0 | |
313.221 | |
JFLSOKIMYBSASW-UHFFFAOYSA-N | |
94524 | |
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3Cl)Cl |
C19H14Cl2 | |
MFCD00040399 | |
2-chlorotrityl chloride, chloro 2-chlorophenyl methylene dibenzene, 1-chloro-2-chlorodiphenylmethyl benzene, 2-chlorophenyldiphenylmethyl chloride, 2-chlorotrityl chloride resin, 2-chlorophenyl diphenylmethyl chloride, 2-chlorotrityl resin, 2-chlorotritylchloride, chloro 2-chlorophenyl diphenylmethane, 2-chlorotrityl chloride, polymer-bound | |
1-chloro-2-[chloro(diphenyl)methyl]benzene |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only