Learn More
2-Chlorotrityl chloride on polystyrene, 1% cross-linked, 100-200 mesh, 1.0-1.4 mmol/g
CAS: 42074-68-0 | C19H14Cl2 | 313.221 g/mol
$170.71 - $1918.89
Identifiants chimiques
| CAS | 42074-68-0 |
|---|---|
| Molecular Formula | C19H14Cl2 |
| Molecular Weight (g/mol) | 313.221 |
| MDL Number | MFCD00040399 |
| InChI Key | JFLSOKIMYBSASW-UHFFFAOYSA-N |
| Synonym | 2-chlorotrityl chloride, chloro 2-chlorophenyl methylene dibenzene, 1-chloro-2-chlorodiphenylmethyl benzene, 2-chlorophenyldiphenylmethyl chloride, 2-chlorotrityl chloride resin, 2-chlorophenyl diphenylmethyl chloride, 2-chlorotrityl resin, 2-chlorotritylchloride, chloro 2-chlorophenyl diphenylmethane, 2-chlorotrityl chloride, polymer-bound |
| PubChem CID | 94524 |
| IUPAC Name | 1-chloro-2-[chloro(diphenyl)methyl]benzene |
| SMILES | C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3Cl)Cl |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
AA4440704
|
Thermo Scientific Chemicals
04440704 |
2 g |
chaque for $170.71
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
AA4440709
|
Thermo Scientific Chemicals
04440709 |
10 g |
chaque for $623.15
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
AA4440718
|
Thermo Scientific Chemicals
04440718 |
50 g |
chaque for $1,918.89
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
2-Chlorotrityl chloride is used as an intermediate of clotrimazole. It is also used in convergent peptide synthesis. Furthermore, it is used in the preparation of modified trityl nucleosides. In addition, it serves as a protecting group reagent for carboxylic acids forming esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 42074-68-0 | |
| 313.221 | |
| JFLSOKIMYBSASW-UHFFFAOYSA-N | |
| 94524 | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3Cl)Cl |
| C19H14Cl2 | |
| MFCD00040399 | |
| 2-chlorotrityl chloride, chloro 2-chlorophenyl methylene dibenzene, 1-chloro-2-chlorodiphenylmethyl benzene, 2-chlorophenyldiphenylmethyl chloride, 2-chlorotrityl chloride resin, 2-chlorophenyl diphenylmethyl chloride, 2-chlorotrityl resin, 2-chlorotritylchloride, chloro 2-chlorophenyl diphenylmethane, 2-chlorotrityl chloride, polymer-bound | |
| 1-chloro-2-[chloro(diphenyl)methyl]benzene |
Spécifications
| Powder | |
| C19H14Cl2 | |
| 2-chlorotrityl chloride, chloro 2-chlorophenyl methylene dibenzene, 1-chloro-2-chlorodiphenylmethyl benzene, 2-chlorophenyldiphenylmethyl chloride, 2-chlorotrityl chloride resin, 2-chlorophenyl diphenylmethyl chloride, 2-chlorotrityl resin, 2-chlorotritylchloride, chloro 2-chlorophenyl diphenylmethane, 2-chlorotrityl chloride, polymer-bound | |
| JFLSOKIMYBSASW-UHFFFAOYSA-N | |
| 1-chloro-2-[chloro(diphenyl)methyl]benzene | |
| 94524 | |
| 2-Chlorotrityl chloride on polystyrene, 1% cross-linked, 1.0 to 1.4 mmol/g |
| 42074-68-0 | |
| MFCD00040399 | |
| 2 g | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3Cl)Cl | |
| 313.221 | |
| 100 to 200 Mesh |
Sécurité et manipulation
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only