missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzoic acid, 98%

CAS: 118-91-2 | C7H5ClO2 | 156.57 g/mol

Supplier:  Thermo Scientific Chemicals 159241000

 View more versions of this product

Catalog No. AC159241000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
    • Recyclable
    • Non-Toxic
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clo

GHS Signal Word: Warning

EINECSNumber : 204-285-4

RTECSNumber : DG4976000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only