missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorobenzaldehyde 99.0+%, TCI Americaâ„¢
$30.24 - $92.60
Chemical Identifiers
CAS | 89-98-5 |
---|---|
Molecular Formula | C7H5ClO |
Molecular Weight (g/mol) | 140.566 |
MDL Number | MFCD00003304 |
InChI Key | FPYUJUBAXZAQNL-UHFFFAOYSA-N |
Synonym | o-chlorobenzaldehyde, chlorobenzaldehyde, benzaldehyde, 2-chloro, benzaldehyde, o-chloro, 2-chlorbenzaldehyd, 2-clorobenzaldeide, o-chloorbenzaldehyde, usaf m-7, 2-chloorbenzaldehyde, o-chlorobenzenecarboxaldehydeShow More |
PubChem CID | 6996 |
IUPAC Name | 2-chlorobenzaldehyde |
SMILES | C1=CC=C(C(=C1)C=O)Cl |
Chemical Identifiers
89-98-5 | |
140.566 | |
FPYUJUBAXZAQNL-UHFFFAOYSA-N | |
6996 | |
C1=CC=C(C(=C1)C=O)Cl |
C7H5ClO | |
MFCD00003304 | |
o-chlorobenzaldehyde, chlorobenzaldehyde, benzaldehyde, 2-chloro, benzaldehyde, o-chloro, 2-chlorbenzaldehyd, 2-clorobenzaldeide, o-chloorbenzaldehyde, usaf m-7, 2-chloorbenzaldehyde, o-chlorobenzenecarboxaldehydeShow More | |
2-chlorobenzaldehyde |
Specifications
89-98-5 | |
212°C | |
MFCD00003304 | |
3265 | |
FPYUJUBAXZAQNL-UHFFFAOYSA-N | |
2-chlorobenzaldehyde | |
6996 | |
≥99.0% (GC) | |
2-Chlorobenzaldehyde |
Yellow | |
C7H5ClO | |
25 g | |
o-chlorobenzaldehyde, chlorobenzaldehyde, benzaldehyde, 2-chloro, benzaldehyde, o-chloro, 2-chlorbenzaldehyd, 2-clorobenzaldeide, o-chloorbenzaldehyde, usaf m-7, 2-chloorbenzaldehyde, o-chlorobenzenecarboxaldehyde | |
C1=CC=C(C(=C1)C=O)Cl | |
140.566 | |
140.57 | |
Liquid |
Safety and Handling
EINECSNumber : (3)-1162
RTECSNumber : CU5075000
TSCA : Yes