Learn More
2-Aminophenol, 99%
CAS: 95-55-6 | C6H7NO | 109.13 g/mol
$60.85 - $72.90
Chemical Identifiers
CAS | 95-55-6 |
---|---|
Molecular Formula | C6H7NO |
Molecular Weight (g/mol) | 109.13 |
MDL Number | MFCD00007690 |
InChI Key | CDAWCLOXVUBKRW-UHFFFAOYSA-N |
Synonym | o-aminophenol, o-hydroxyaniline, 2-hydroxyaniline, phenol, 2-amino, 2-amino-1-hydroxybenzene, fouramine op, benzofur gg, pelagol grey gg, pelagol 3ga, nako yellow 3gaShow More |
PubChem CID | 5801 |
ChEBI | CHEBI:18112 |
IUPAC Name | 2-aminophenol |
SMILES | C1=CC=C(C(=C1)N)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC104260050
|
Thermo Scientific Chemicals
104260050 |
5 g | Glass bottle |
Each for $60.85
|
|
|||||
AC104261000
|
Thermo Scientific Chemicals
104261000 |
100 g | Glass bottle |
Each for $72.90
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
95-55-6 | |
109.13 | |
CDAWCLOXVUBKRW-UHFFFAOYSA-N | |
5801 | |
2-aminophenol |
C6H7NO | |
MFCD00007690 | |
o-aminophenol, o-hydroxyaniline, 2-hydroxyaniline, phenol, 2-amino, 2-amino-1-hydroxybenzene, fouramine op, benzofur gg, pelagol grey gg, pelagol 3ga, nako yellow 3gaShow More | |
CHEBI:18112 | |
C1=CC=C(C(=C1)N)O |
Specifications
95-55-6 | |
100.0 | |
White to Brown | |
168°C | |
98.5% min. (GC) | |
C6H7NO | |
MFCD00007690 | |
13, 354 | |
o-aminophenol, o-hydroxyaniline, 2-hydroxyaniline, phenol, 2-amino, 2-amino-1-hydroxybenzene, fouramine op, benzofur gg, pelagol grey gg, pelagol 3ga, nako yellow 3ga | |
CDAWCLOXVUBKRW-UHFFFAOYSA-N | |
2-aminophenol | |
5801 | |
109.13 | |
Powder |
98.5 | |
172°C to 177°C | |
164°C (15.0 Mbar) | |
Authentic | |
Glass bottle | |
H2NC6H4OH | |
5 g | |
15, 457 | |
Solubility in water: 17g/L (20°C). Other solubilities: soluble in ether,1g in 23 mL alcohol,slightly soluble in benzene | |
C1=CC=C(C(=C1)N)O | |
109.13 | |
CHEBI:18112 | |
99% | |
2-Aminophenol, 99% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN:
GHS Signal Word: Warning
EINECSNumber : 202-431-1
RTECSNumber : SJ4950000
TSCA : TSCA
RUO – Research Use Only